Would you like to log in again?
BLOWER FOR 30192-705
Supplier: Labconco
Remote Blower^ Designed for use with Protector* workstation that needs remote blower. Has 1/4hp direct-drive motor. Housing and wheel are corrosion-resistant enamel-coated steel. Discharge direction is adjustable. Inlet is 15.2cm I.D.; outlet is 18.7O.D.x10.8Dcm. 115V, 60Hz.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BLOWER FOR 30192-705 30192-706 7053500 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty