Would you like to log in again?
DRYING TRAIN F/CA GLV BOX 230V
Supplier: Labconco
Drying train for ca glove box 230v^ 220v 50/60 Hz. Drying train provides a means for reducing the humidity within the glove box to less than 5 ppm. includes two molecular sieve columns, 1 cf. pump and stainless steel tubing to connect column to pump. Glove box accessory.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
DRYING TRAIN F/CA GLV BOX 230V 32940-055 5061303 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty