Would you like to log in again?
CentriVap® Centrifugal Concentration System Accessories, Labconco®
Supplier: Labconco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CentriVap®
7397605 7450700 7824700 7455000 7455101 7814500 7553100 7815000 7456600 7553501 7814900 7397700 7461901 7455100 7991000 7814300 7455200 7815200 7462901 7455001 7461900 7995600 7464300 7460900 7814800 7462900 7824702 7553500 7450701 7455201 7474000 7815300 7914101
37000-596EA 1368.38 USD InStock
37000-596 89259-754 89041-252 26676-010 89239-682 37001-398 37001-396 26617-040 37001-394 37001-392 26675-877 37001-390 15398-028 26675-878 26675-879 89041-256 89041-254 37000-590 89004-964 89004-966 89041-258 89041-260 74920-006 89041-262 37001-400 37001-388 26675-885 26675-881 75779-944 83009-292 89259-756 75779-942 26675-884
CentriVap® Centrifugal Concentration System Accessories, Labconco®
Concentrators
For use with all CentriVap® centrifugal concentration systems, except DNA system.
CentriZap® strobe light enables user to shine a strobe light into the chamber to easily view samples while the rotor is spinning. Samples appear to be standing still. Includes a holster bracket (with mounting hardware) that attaches to the right side of the unit. Glass lid provides additional protection from solvents and corrosives that attack acrylic. Vacuum controller provides manual control and monitoring of vacuum levels. Analog gauge shows vacuum from 0–762 mm Hg (0–30" Hg). Three-way valve is polypropylene and has an O.D. of 1.3 cm (½"). Bleed valve bleeds vacuum manually when using cold trap with other laboratory equipment. Vacuum tubing kit includes two 1.3 cm (½") I.D. vacuum tubing pieces, 121.9 cm (48") in length, and four tubing clamps. Two-port manifold allows freeze drying of small volume samples in freeze dry flasks, serum bottles, or ampules using a CentriVap® system. Glass trap insert protects cold trap reservoir and lid from corrosives. Clear polypropylene canister with mounting bracket attaches to the side of the cold trap. Includes 43.2 cm (17") PVC tubing and two clamps. Canister with stand recommended when more than one chemical trap is required.
Accessory rotors are constructed of anodized aluminum and hold tubes at an angle to maximize surface area. Rotors are available standard, or with a fluoropolymer resin coating that ensures resistance to acids. The 12–13 mm rotor holds a maximum of 148 tubes: (40) 1.5 to 2.0 ml microcentrifuge tubes and up to (108) 12 to 13 mm tubes. The 12 to 17 mm rotor holds a maximum of 76 tubes: up to (52) 12 to 13 mm tubes and up to (24) 16 to 17 mm tubes. The 28 mm rotor holds 44 tubes: (32) 1.5 to 2.0 ml microcentrifuge tubes and (12) 28 mm tubes. Hexagonal rotor holds (132) 1.5 to 2.0 ml microcentrifuge tubes. The DNA rotor holds a maximum of 132 tubes: up to (72) 0.5 ml and up to (60) 1.5 to 2.0 ml microcentrifuge tubes. Microtiter plate rotor holds four standard or two deep well plates. The High Capacity Micro Rotor holds up to 148 micro centrifuge tubes, 1.5 or 2.0 ml, and micro auto sample vials approx. 12 mm in dia. ×35 mm. The High Capacity Micro Rotor doesn't require a lip on the tube and is perfect for vials.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty