Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

RapidVap® N₂/48 Evaporation Systems, 230 V

Supplier: Labconco
RapidVap®
79100-13 79100-15
37001-582EA 21662.51 USD InStock
37001-582 37001-578
RapidVap® N₂/48 Evaporation Systems, 230 V
Evaporators Centrifugal Evaporators
Evaporation systems use nitrogen blow down, vortex motion, and heat to speed up evaporation of up to 48 small samples or eight 450 ml samples at once

  • 230 V model

Systems employ eight clusters of six ports each, and nitrogen may be delivered to 2, 4, 6, or 8 clusters at once so that nitrogen is conserved when a full load is not required. Microprocessor-controlled vortex motion (up to 500 rpm) increases surface area for faster evaporation while continually mixing the sample to maximize recovery. Utilizing a dry block heating method (up to 100 °C), systems provide a more reliable alternative to maintenance-intensive, contamination-prone water baths. Cabinet base is constructed of epoxy-coated cast aluminum. The upper housing is solvent-resistant, smooth acrylic/PVC thermoplastic. Chemical-resistant glass lid provides visibility of samples, and two clamps hold the lid securely in place. PTFE-coated aluminum chamber conducts heat efficiently and resists corrosion. Aluminum sample blocks are PTFE-coated.

Optional two-way RS-232 link allows remote control of functions from a user-supplied personal computer.

A stream of nitrogen or dry gas is directed downward onto the surface of the sample. Nitrogen blow down reduces the partial pressure directly over the liquid to speed evaporation and help remove the solvent as it evaporates. Systems may be used to prepare samples for applications such as drug development and metabolism, bioresearch, combinatorial chemistry, environmental testing, forensics, and toxicology.

Caution: Systems should be located within a fume hood if hazardous or flammable solvents are used. In all cases, regardless of the solvent used, it is recommended that the exhaust hose be vented into a fume hood or other laboratory ventilation device.

Note: Actual sample volumes of tubes depend on tube shape, height, and vortex speed.

Certifications: 230V models are CE marked.

Ordering information: One-year manufacturer's warranty. Required accessories include an aluminum sample block (sold separately), glassware, nitrogen source with a maximum flow rate of 3.5CFM, and a gas pressure regulator. For glassware, nitrogen, and gas pressure regulator, contact your Avantor representative.

Caution: 230 V model. Corresponding models with voltage suitable for use in US (37001-576 and 37001-580) are also available, please see separate product page.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty