Would you like to log in again?
Solid Epoxy Flat Work Surfaces, Labconco®
Supplier: Labconco
4882805 4882805 4882803 4882804 4882803 4882806 4882806 4882804
470203-230EA 1384.99 USD InStock
470203-230 470203-224 46610-522 82001-230 470203-228 470203-226 46610-490 46610-492
Solid Epoxy Flat Work Surfaces, Labconco®
Fume Hoods and Enclosures Chemical Fume Hoods
Combine with a base stand or cabinet to support Fiberglass 30 and Basic™ Labconco® laboratory hoods.
  • For Use with Base Stands and Cabinets
  • Designed to Support Fiberglass 30 and Basic Labconco Laboratory Hoods
  • Black Surfaces
  • Depth: 76.2 cm (30")
  • Thickness: 3.2 cm (1¼")
Surfaces are black.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty