Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Solid Epoxy Flat Work Surfaces, Labconco®

Supplier: Labconco
4882805 4882805 4882803 4882804 4882803 4882806 4882806 4882804
470203-230EA 1384.99 USD InStock
470203-230 470203-224 46610-522 82001-230 470203-228 470203-226 46610-490 46610-492
Solid Epoxy Flat Work Surfaces, Labconco®
Fume Hoods and Enclosures Chemical Fume Hoods
Combine with a base stand or cabinet to support Fiberglass 30 and Basic™ Labconco® laboratory hoods.

  • For Use with Base Stands and Cabinets
  • Designed to Support Fiberglass 30 and Basic Labconco Laboratory Hoods
  • Black Surfaces
  • Depth: 76.2 cm (30")
  • Thickness: 3.2 cm (1¼")

Surfaces are black.

They have a depth of 76.2 cm (30") and a width of 3.2 cm (11/4").

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty