Would you like to log in again?
UV LAMP KIT FOR PURIFIER DELTA
Supplier: Labconco
UV Lamp Kit^ For the 1.8m Purifier* Delta* Series Class II Type A2 and B2 safety cabinets. Includes a 254nm UV lamp and installation instructions.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
UV LAMP KIT FOR PURIFIER DELTA 46610-774 3745001 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty