Would you like to log in again?
TOP RACK REPL ADJST HGT SS
Supplier: Labconco
Adjustable Height Top Rack^ For SteamScrubber* Washer. Stainless steel with acetal rollers. Expanded versatility in handling glassware. Has two 24-pin insert baskets that may be positioned high or low. Pins have plastic tips to protect glassware. Use in place of standard top rack.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TOP RACK REPL ADJST HGT SS 66074-915 4494501 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty