Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 37% GB for the electronics industry

Supplier: KMG
Synonyms: Muriatic acid

64011
71009-358CS 980.17 USD InStock
71009-358
Hydrochloric acid 37% GB for the electronics industry
Hydrochloric acid
Hydrochloric Acid is a key ingredient to remove surface metallic ions by forming soluble metal chlorides.


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Assay (HCl) 37.00 - 38.00%
Color 10 APHA
Appearance Conform
Phosphate 50 ppb
Sulfate 200 ppb
Sulfite 700 ppb
Aluminum 5 ppb
Antimony 1 ppb
Arsenic 1 ppb
Barium 1 ppb
Boron 10 ppb
Cadmium 1 ppb
Calcium 10 ppb
Chromium 2 ppb
Cobalt 1 ppb
Copper 1 ppb
Gallium 1 ppb
Germanium 1 ppb
Gold 1 ppb
Iron 10 ppb
Lead 1 ppb
Lithium 1 ppb
Magnesium 1 ppb
Manganese 1 ppb
Molybdenum 1 ppb
Nickel 1 ppb
Niobium 1 ppb
Potassium 3 ppb
Silver 1 ppb
Sodium 3 ppb
Strontium 1 ppb
Tin 5 ppb
Titanium 1 ppb
Vanadium 1 ppb
Zinc 5 ppb
0.50µ Particle Count 50 par/ml
1.00µ Particle Count 10 par/ml

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty