Would you like to log in again?
KIT WATERPRO MOBILE/HLPC
Supplier: Labconco
Kit, Waterpro mobile/HPLC^ HPLC polishing kit for Waterpro mobile system 90901-00 and 90901-01 (manufactured before 8/1/93). includes 1 prefilter, 1 carbon/prefilter, 1 di cartridge, 1 organic adsorption cartridge and 2 hollow fiber final filters. Water purif. supplies.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
KIT WATERPRO MOBILE/HLPC 72450-012 9092400 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty