Would you like to log in again?
FILTER FOR MCLEOD GAUGE
Supplier: Labconco
Filter for mcleod gauge^ Borosilicate glass. Filter protects mcleod gauge from dust and other contaminants. borosilicate glass bulb filled with fiberglass has tube connections at both ends. uses 5/16" id x 3/16" wall rubber tubing for connection to gauge. Freeze dry accessory.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FILTER FOR MCLEOD GAUGE 72460-016 7585100 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty