Would you like to log in again?
ADAPTER AMPULE 3-WAY
Supplier: Labconco
Adapter ampule 3-way^ Borosilicate glass. Trident (3-way) adapter permits attachment of three ampules to a single valve port. ampules are attached using ampule valve adapters. cavity in adapter body can be filled with cotton fiber media to help prevent contamination between samples.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER AMPULE 3-WAY 72460-054 7762700 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty