Would you like to log in again?
RapidVap® Trapping Valve, Labconco®
Supplier: Labconco
7348000
75841-004EA 1863.1 USD InStock
75841-004
RapidVap® Trapping Valve, Labconco®
Valves Vacuum Valves
The RapidVap® Trapping Valve aids in solvent recovery for RapidVap® Vacuum Evaporation Systems.
  • Cold Trap is required (not included)
  • Includes tubing and clamps for connection to Vacuum System
  • Compatible with RapidVap® Vacuum Systems manufactured after September 2016
Ordering information: RapidVap® Vacuum System and Cold Trap each sold separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty