Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
FreeZone® 2.5 L Benchtop Freeze Dryers, Labconco®
Supplier: Labconco
FreeZone®
700201010 710201040 710202015 710202040 700202040 710201010 700201070 700201000 710202050 710202010 710201030 700202030 710202000 700201040 700202050 700201050 710201070 710202030 700201030 700202000 710201000 710201015 700202070 710201050 700202015 700201015 700202010 710202070
75999-782EA 11278.61 USD InStock
75999-782 75999-780 75999-796 76000-460 75999-794 76000-462 75999-778 75999-812 76000-464 75999-798 75999-810 76000-466 75999-792 75999-790 75999-786 75999-784 75999-800 76000-452 75999-788 76000-454 75999-804 76000-456 76000-632 75999-802 76000-458 76000-634 75999-808 75999-806
FreeZone® 2.5 L Benchtop Freeze Dryers, Labconco®
Freeze Dryers
FreeZone 2.5 Liter Benchtop Freeze Dryers are designed for lyophilizing light sample loads. The compact design is ideal for labs with limited floor space. Collectors are available to -50° C or -84° C.
  • Stay close to your samples as Lyo-Works™ operating system, provides mobile connectivity
  • Consistent, high quality results with flexible programming, and on board data collection
  • Challenging samples are easy with accessories designed specifically for laboratory samples
  • End-Zone™ End Point Detection option takes the guess work out of sample dryness
New Lyo-Works™ OS provides flexibility and convenience in an easy-to-use, touch screen display
The upright stainless steel collector coil holds 2.5 liters of ice before defrosting. Choice of a -50° C (for aqueous samples) or -84° C (for low eutectic point samples including acetonitrile) collector cooled by an environmentally-friendly HCFC/CFC-free refrigeration system. Internal coil maximizes vapor-trapping efficiency. Optional PTFE-coated collectors withstand corrosive compounds.
Lyo-Works OS provides a real time display of collector temperature and vacuum level on a 5" full color touch screen. Flexible programming and control of optional drying accessories including End-Zone™ end point detection for flask freeze drying. One-touch data logging and display provides information to maximize sample quality and minimize run time. Customizable settings include language, time zone, temperature and vacuum. Alerts and feedback of current operating parameters can be sent automatically to user-entered email addresses.
FreeZone exclusive features protect your vacuum pump from costly damage. A vacuum break valve bleeds air into the system when power to the freeze dryer or vacuum pump is shut off for more than 5 minutes. A moisture sensor prevents refrigeration or vacuum start-up when moisture is detected in the drain line. Vacuum pump is automatically shut down when a leak is present in initial pull down or during run.
Certifications: 60 Hz models conform to UL Standard 61010-1 and CAN/CSA C22.2 No. 61010-1. 50 Hz models conform to CE Conformity marking.
Accessories information: Stainless steel lids are optional collector lids for FreeZone Freeze Dryers. They have 10.1" dia., with neoprene gasket and 3" dia. hole for direct connection of drying accessories to collector.
Ordering information: Vacuum pump with a displacement of at least 98 L/min., drying accessory and freeze dry glassware (if not bulk freeze drying) are required (not included).
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty