Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

FreeZone® Benchtop Freeze Dryers, 8 L, Labconco®

Supplier: Labconco
FreeZone®
700802000 700801015 700801000 700802015 700802070 700802030 700802040 700802010 700801040 700801070 700801050 700801030 700802050 700801010
76000-508EA 18394.19 USD InStock
76000-508 75999-862 75999-852 75999-860 75999-850 75999-844 75999-856 75999-842 75999-864 75999-854 75999-848 76000-510 75999-846 75999-858
FreeZone® Benchtop Freeze Dryers, 8 L, Labconco®
Freeze Dryers
FreeZone 8 Liter Benchtop Freeze Dryers are designed for lyophilizing moderate sample loads. The compact design is ideal for labs with limited floor space.

  • Stay close to your samples as Lyo-Works™ operating system, provides mobile connectivity
  • Consistent, high quality results with flexible programming, and on board data collection
  • Challenging samples are easy with accessories designed specifically for laboratory samples
  • End-Zone™ End Point Detection option takes the guess work out of sample dryness

New Lyo-Works™ OS provides flexibility and convenience in an easy-to-use, touch screen display

The upright stainless steel collector coil holds 8 liters of ice before defrosting. The -50° C (for aqueous samples) collector is cooled by an environmentally-friendly HCFC/CFC-free refrigeration system. Internal coil maximizes vapor-trapping efficiency. Optional PTFE-coated collectors withstand corrosive compounds.

Lyo-Works OS provides a real time display of collector temperature and vacuum level on a 5"" full color touch screen. Flexible programming and control of optional drying accessories including End-Zone™ end point detection for flask freeze drying. One-touch data logging and display provides information to maximize sample quality and minimize run time. Customizable settings include language, time zone, temperature and vacuum. Alerts and feedback of current operating parameters can be sent automatically to user-entered email addresses.

FreeZone exclusive features protect your vacuum pump from costly damage. A vacuum break valve bleeds air into the system when power to the freeze dryer or vacuum pump is shut off for more than 5 minutes. A moisture sensor prevents refrigeration or vacuum start-up when moisture is detected in the drain line. Vacuum pump is automatically shut down when a leak is present in initial pull down or during run.

Certifications: 60 Hz models conform to UL Standard 61010-1 and CAN/CSA C22.2 No. 61010-1. 50 Hz models conform to CE Conformity marking.

Accessories information: Acrylic lid have 14.0" dia., with neoprene gasket and 3" dia. hole for direct connection of drying accessories to
collector. Includes cap for 3" dia. attachment port.

Ordering information: Vacuum pump with a displacement of at least 98 liters/minute, drying accessory and freeze dry glassware (if not bulk freeze drying) are required (not included).

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty