Would you like to log in again?
FreeZone® 18 L Console Freeze Dryers, Labconco®
Supplier: Labconco
FreeZone®
701812250 701811240 701811030 701812140 701812040 701811050 701812110 701811230 701812210 701812215 701812270 701811210 701811040 701812030 701812115 701811010 701812240 701811170 701812130 701811130 701812170 701811215 701812230 701811115 701811015 701812050 701812070 701812015 701811140 701811250 701811110 701811070 701812150 701812010 701811270 701811150
76000-180EA 37762.03 USD InStock
76000-180 76000-182 76000-160 76000-184 76000-162 76000-140 76000-164 76000-560 76000-186 76000-142 76000-188 76000-144 76000-562 76000-166 76000-146 76000-168 76000-564 76000-148 76000-566 76000-190 76000-192 76000-170 76000-172 76000-150 76000-174 76000-152 76000-176 76000-154 76000-552 76000-156 76000-178 76000-158 76000-554 76000-556 76000-138 76000-558
FreeZone® 18 L Console Freeze Dryers, Labconco®
Freeze Dryers
FreeZone® 18 Liter Console Freeze Dryers are designed for lyophilizing moderate to large sample loads or numerous small container batches.
  • Stay close to your samples as Lyo-Works™ operating system, provides mobile connectivity
  • Consistent, high quality results with flexible programming, and on board data collection
  • Challenging samples are easy with accessories designed specifically for laboratory samples
  • End-Zone™ End Point Detection option takes the guess work out of sample dryness
New Lyo-Works™ OS provides flexibility and convenience in an easy-to-use, touch screen display
The upright stainless steel collector coil holds 18 liters of ice before defrosting. The -50° C (for aqueous samples) collector is cooled by an environmentally-friendly HCFC/CFC-free refrigeration system. Internal coil maximizes vapor-trapping efficiency. Optional PTFE-coated collectors withstand corrosive compounds.
Lyo-Works OS provides a real time display of collector temperature and vacuum level on a 5"" full color touch screen. Flexible programming and control of optional drying accessories including End-Zone™ end point detection for flask freeze drying. One-touch data logging and display provides information to maximize sample quality and minimize run time. Customizable settings include language, time zone, temperature and vacuum. Alerts and feedback of current operating parameters can be sent automatically to user-entered email addresses.
FreeZone exclusive features protect your vacuum pump from costly damage. A vacuum break valve bleeds air into the system when power to the freeze dryer or vacuum pump is shut off for more than 5 minutes. A moisture sensor prevents refrigeration or vacuum start-up when moisture is detected in the drain line. Vacuum pump is automatically shut down when a leak is present in initial pull down or during run. Front viewing window makes monitoring vacuum pump oil hassle free.
Your samples are protected with remote notification and data recording. Features include hot gas defrost, side-mounted USB port and rear-mounted Ethernet connection. The console provides interior space for a vacuum pump and accommodates the FreeZone Stoppering Tray Dryer (pump and tray dryer sold separately).
Certifications: 60 Hz models conform to UL Standard 61010-1 and CAN/CSA C22.2 No. 61010-1. 50 Hz models conform to CE Conformity marking.
Accessories information: Stainless steel lid have 14.0" dia., with neoprene gasket and 3" dia. hole for direct connection of drying accessories to collector.
Includes cap for 3" dia. attachment port. Acrylic lid have 14.0" dia., with neoprene gasket and 3" dia. hole for direct connection of drying accessories to collector. Includes cap for 3" dia. attachment port.
Ordering information: Vacuum pump with a displacement of at least 98 liters/minute, drying accessory and freeze dry glassware (if not bulk freeze drying) are required (not included).
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty