Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
FreeZone® Stoppering Tray Dryers, Labconco®
Supplier: Labconco
794801015 794801340 794801250 794801150 794801370 794801300 794801270 794801130 794801170 794801000 794801140 794801040 794801315 794801310 794801050 794801240 794801030 794801210 794801215 794801115 794801350 794801070 794801110 794801330 794801100 794801230 794801200 794801010
76000-380EA 44088.15 USD InStock
76000-380 76000-382 76000-360 76000-384 76000-362 76000-386 76000-364 76000-388 76000-366 76000-346 76000-368 76000-348 76000-624 76000-626 76000-390 76000-370 76000-392 76000-350 76000-372 76000-394 76000-396 76000-352 76000-374 76000-354 76000-376 76000-356 76000-378 76000-358
FreeZone® Stoppering Tray Dryers, Labconco®
Freeze Dryers
FreeZone Stoppering Tray Dryers provide precise control and program flexibility for consistent performance and are designed for lyophilizing moderate to large loads and stoppering vials or serum bottles under vacuum. They offer options for a built-in 6-port manifold and isolation valve.
  • Compact design for moderate to large sample loads or numerous sample batches
  • Precision with 3 temperature-controlled stoppering shelves between -40 and +40 °C
  • Flexibility with temperature and vacuum level control
  • Quality reassurance with onboard data collection, and real time graphing
  • Sample probes with remote notification of operating parameters or alerts
The chamber pre-freezes samples to save time and money by eliminating the need for a separate freezer and product transfer
Three 16.5" x 12" temperature-controlled (±1 °C), adjustable shelves with sample temperature sensors provide up to 594 sq. inches of shelf space. Circulating fluid in shelves ranges from -40 and +40 °C and features an environmentally-friendly HCFC/CFC-free refrigeration system. A clear acrylic viewing door provides complete visibility of the shelves. The 1/8" OD backfill port introduces sterile air or inert gas from an outside source to the chamber, protecting samples from atmospheric moisture and contaminants.
Lyo-Works™ OS provides a real time display of freeze dryer and tray dryer parameters on a 5" full color touch screen on the connected FreeZone 6, 12, or 18 Liter Freeze Dryer (sold separately). Data may be exported via USB or Ethernet. A microprocessor-based controller allows up to 30 different temperature programs to be stored and repeated, each using up to 16 different steps. The display provides navigation to manual or program mode, probe temperatures, vacuum in mBar, Torr or Pa, step of the program in progress, time remaining in present step, and end of program, as well as alarms. Alerts and current operating parameter reading can be sent automatically to user-entered email addresses.
Optional 6-Port Tray Dryer Manifold accommodates either 1/2" or 3/4" flask adapters and connect to flasks and other glassware. The optional built-in isolation valve separates the Stoppering Tray Dryer from the freeze dryer to shorten time necessary to reestablish working vacuum levels and to provide a means for determining end point.
Certifications: 60 Hz models conform to UL Standard 61010-1 and CAN/CSA C22.2 No. 61010-1. 50 Hz models conform to CE Conformity marking.
Accessories information: Stainless Steel Bulk Tray, holds liquids for bulk drying.
Ordering information: FreeZone 6, 12, or 18 Liter Freeze Dryer with Lyo-Works OS, vacuum pump with displacement of at least 163 liters/minute, and freeze dry glassware (if not bulk freeze drying) are required (not included).
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty