Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

FreeZone® Small Tray Dryers, Labconco®

Supplier: Labconco
FreeZone®
780701040 780701050 780701015 780701030 780701000 780701070 780701010
76000-450EA 13304.86 USD InStock
76000-450 76000-440 76000-442 76000-444 76000-446 76000-448 76000-438
FreeZone® Small Tray Dryers, Labconco®
Freeze Dryers
FreeZone Small Tray Dryers provide precise control and program flexibility for consistent performance and are designed for lyophilizing small to moderate bulk sample loads or numerous sample batches.

  • Compact design for small bulk loads or numerous sample batches
  • Precision with 3 heated shelves up to +60°C
  • Flexibility: temperature and vacuum level controls in 30 programs with up to 16 segments each
  • Quality reassurance with onboard data collection and real time graphing
  • Sample probes with remote notification of operating parameters or alerts

Three 10.6” x 11.7” heated shelves with sensors for monitoring sample temperature provide 372 sq

Lyo-Works™ OS provides flexible programming and control with a real time display of freeze dryer and tray dryer parameters on a 5"" full color touch screen on the connected FreeZone Freeze Dryer (sold separately). One-touch data logging and display provides information to maximize sample quality and minimize run time. Customizable settings include language, time zone, temperature and vacuum. Alerts and feedback of current operating parameters can be sent automatically to user-entered email addresses.

Samples must be pre-frozen.

inches of shelf space with the option for 2 additional shelves (sold separately). Each shelf warms to +60°C and a microprocessor maintains the system temperature within 3°C of set point. A clear acrylic viewing door provides complete visibility of the shelves.

Certifications: 60 Hz models conform to UL Standard 61010-1 and CAN/CSA C22.2 No. 61010-1. 50 Hz models conform to CE Conformity marking.

Ordering information: FreeZone Freeze Dryer with Lyo-Works OS and a vacuum pump with displacement of at least 98 liters/minute are required (not included).

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty