Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
FreeZone® 6 L Console Freeze Dryers with Stoppering Tray Dryer, Labconco®
Supplier: Labconco
FreeZone®
700621140 710621040 700622015 700621050 700622030 710622110 710622150 700622140 710622115 700622100 700622000 710622130 700621110 700621070 700621010 710622015 700622130 710621150 700621040 700622040 710622040 710622140 700622170 710621050 710622170 700621030 700622150 700621000 710621170 710622070 710622100 710621010 700622070 710621100 710621015 700622115 710621000 710621070 710622050 700621150 700621115 700622050 710622010 710621110 710621030 710622030 700621130 700622110 710622000 700622010 700621015 700621100 710621130 710621115 710621140 700621170
76000-260EA 65592.77 USD InStock
76000-260 76000-262 76000-264 76000-220 76000-222 76000-266 76000-224 76000-268 76000-226 76000-228 76000-270 76000-194 76000-272 76000-196 76000-274 76000-230 76000-198 76000-672 76000-276 76000-232 76000-278 76000-674 76000-234 76000-676 76000-236 76000-238 76000-678 76000-280 76000-240 76000-680 76000-242 76000-684 76000-200 76000-244 76000-202 76000-686 76000-246 76000-204 76000-248 76000-688 76000-568 76000-206 76000-208 76000-250 76000-570 76000-252 76000-572 76000-210 76000-254 76000-574 76000-212 76000-256 76000-258 76000-214 76000-216 76000-218
FreeZone® 6 L Console Freeze Dryers with Stoppering Tray Dryer, Labconco®
Freeze Dryers
FreeZone 6 Liter Console Freeze Dryers with Stoppering Tray Dryers are designed for lyophilizing moderate to large loads and stoppering vials under vacuum. Collectors are available to -50° C or -84° C.
  • Stay close to your samples as Lyo-Works™ operating system, provides mobile connectivity
  • Consistent, high quality results with flexible programming, and on board data collection
  • Challenging samples are easy with accessories designed specifically for laboratory samples
  • End-Zone™ End Point Detection option takes the guess work out of sample dryness
New Lyo-Works™ OS provides flexibility and convenience in an easy-to-use, touch screen display
The upright stainless steel collector coil holds 6 liters of ice before defrosting. Choice of a -50° C (for aqueous samples) or -84° C (for low eutectic point samples including acetonitrile) collector cooled by an environmentally-friendly HCFC/CFC-free refrigeration system. Internal coil maximizes vapor-trapping efficiency. Optional PTFE-coated collectors withstand corrosive compounds.
Lyo-Works OS provides a real time display of collector temperature and vacuum level on a 5"" full color touch screen. Flexible programming and control of optional drying accessories including End-Zone™ end point detection for flask freeze drying. One-touch data logging and display provides information to maximize sample quality and minimize run time. Customizable settings include language, time zone, temperature and vacuum. Alerts and feedback of current operating parameters can be sent automatically to user-entered email addresses.
FreeZone exclusive features protect your vacuum pump from costly damage. A vacuum break valve bleeds air into the system when power to the freeze dryer or vacuum pump is shut off for more than 5 minutes. A moisture sensor prevents refrigeration or vacuum start-up when moisture is detected in the drain line. Vacuum pump is automatically shut down when a leak is present in initial pull down or during run. Front viewing window makes monitoring vacuum pump oil hassle free.
Your samples are protected with remote notification and data recording. Features include hot gas defrost, side-mounted USB port and rear-mounted Ethernet connection. The console provides interior space for a vacuum pump (sold separately).
Certifications: 60 Hz models conform to UL Standard 61010-1 and CAN/CSA C22.2 No. 61010-1. 50 Hz models conform to CE Conformity marking.
Accessories information: Stainless steel lid have 14.0" dia., with neoprene gasket and 3" dia. hole for direct connection of drying accessories to collector.
Includes cap for 3" dia. attachment port. Acrylic lid have 14.0" dia., with neoprene gasket and 3" dia. hole for direct connection of drying accessories to collector. Includes cap for 3" dia. attachment port.
Ordering information: Vacuum pump with a displacement of at least 163 liters/minute and freeze dry glassware (if not bulk freeze drying) are required (not included).
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty