Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
FreeZone® 12L Console Freeze Dryers with Stoppering Tray Dryer, Labconco®
Supplier: Labconco
FreeZone®
701222170 711221050 701222010 711222140 701221070 701222030 701222040 701221140 701221010 701222070 711222150 711221140 711222015 701221150 711221170 701222050 701221015 711221010 701221115 711222030 711221150 711222110 701222115 711222130 711221070 701221110 701222140 711221115 711222115 711221015 711222050 701221130 701222015 711222010 701222130 701221170 701221040 711222040 701222150 711222170 701221050 711221040 711222070 711221110 701222110 711221030 701221030 711221130
76000-738EA 71150.5 USD InStock
76000-738 76000-282 76000-580 76000-284 76000-582 76000-286 76000-320 76000-584 76000-288 76000-300 76000-586 76000-740 76000-588 76000-302 76000-600 76000-700 76000-304 76000-602 76000-306 76000-604 76000-308 76000-606 76000-608 76000-290 76000-292 76000-590 76000-690 76000-294 76000-592 76000-296 76000-692 76000-594 76000-694 76000-298 76000-310 76000-596 76000-696 76000-598 76000-576 76000-312 76000-698 76000-610 76000-578 76000-314 76000-316 76000-734 76000-318 76000-736
FreeZone® 12L Console Freeze Dryers with Stoppering Tray Dryer, Labconco®
Freeze Dryers
FreeZone 12 Liter Console Freeze Dryers with Stoppering Tray Dryers are designed for lyophilizing moderate to large loads and stoppering vials or serum bottles under vacuum. Collectors are available to -50° C or -84° C.
  • Stay close to your samples as Lyo-Works™ operating system, provides mobile connectivity
  • Consistent, high quality results with flexible programming, and on board data collection
  • Challenging samples are easy with accessories designed specifically for laboratory samples
  • End-Zone™ End Point Detection option takes the guess work out of sample dryness
New Lyo-Works™ OS provides flexibility and convenience in an easy-to-use, touch screen display
The console’s upright stainless steel collector coil is capable of holding 12 liters of ice before defrosting. Choice of a -50° C (for aqueous samples) or -84° C (for low eutectic point samples including acetonitrile) collector cooled by an environmentally-friendly HCFC/CFC-free refrigeration system. Internal coil maximizes vapor-trapping efficiency. Optional PTFE-coated collectors withstand corrosive compounds.
Lyo-Works OS provides a real time display of collector temperature and vacuum level on a 5"" full color touch screen. Flexible programming and control of optional drying accessories including End-Zone™ end point detection for flask freeze drying. One-touch data logging and display provides information to maximize sample quality and minimize run time. Customizable settings include language, time zone, temperature and vacuum. Alerts and feedback of current operating parameters can be sent automatically to user-entered email addresses.
FreeZone exclusive features protect your vacuum pump from costly damage. A vacuum break valve bleeds air into the system when power to the freeze dryer or vacuum pump is shut off for more than 5 minutes. A moisture sensor prevents refrigeration or vacuum start-up when moisture is detected in the drain line. Vacuum pump is automatically shut down when a leak is present in initial pull down or during run. Front viewing window makes monitoring vacuum pump oil hassle free.
Your samples are protected with remote notification and data recording. Features include hot gas defrost, side-mounted USB port and rear-mounted Ethernet connection. The console provides interior space for a vacuum pump (sold separately).
Accessories information: Stainless steel lid have 14.0" dia., with neoprene gasket and 3" dia. hole for direct connection of drying accessories to collector.
Includes cap for 3" dia. attachment port. Acrylic lid have 14.0" dia., with neoprene gasket and 3" dia. hole for direct connection of drying accessories to
collector. Includes cap for 3" dia. attachment port.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty