Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

FreeZone® 6 L Console Freeze Dryers, Labconco®

Supplier: Labconco
FreeZone®
710611040 700612350 710611230 710611130 710612130 700611070 700612110 710611070 710612100 700611315 700611340 700612330 700612150 700612230 700612130 710611050 700611210 700611230 710611010 710612115 710611210 710612230 700611300 710612030 710611115 700612010 700612100 700611240 710612010 700612200 710611240 700611140 700611000 710611215 700611100 700611350 710612000 710612070 710611140 700612310 700611040 710612215 700612300 700612240 710612240 710612170 710611250 700611370 700611170 700612315 710612150 700612140 700612015 710612110 700611330 710611170 700612210 710612015 700612340 700612050 710612200 700611310 710612140 710611270 700612030 710611015 700611010 710612250 700612070 700612040 700612115 710612040 700611270 710612270 710611030 710611150 700611250 700611050 700611215 710611110 710611000 700612000 700612215 710611200 710611100 700611030 700612370 700611150 700611200 700611115 700611015 700611110 700612250 710612210 700611130 710612050 700612170 700612270
75999-896EA 20488.86 USD InStock
75999-896 75999-894 75999-932 75999-930 75999-898 75999-936 75999-934 75999-938 75999-940 75999-944 75999-942 75999-948 76000-512 75999-946 76000-514 76000-516 76000-518 75999-950 75999-954 76000-000 75999-952 76000-002 76000-520 75999-958 76000-004 76000-522 75999-956 76000-006 76000-524 76000-008 76000-526 76000-528 75999-962 75999-960 75999-966 76000-530 75999-964 76000-652 76000-532 76000-654 76000-534 75999-968 76000-656 76000-536 76000-658 76000-538 75999-980 75999-972 75999-970 75999-976 76000-540 75999-974 76000-542 76000-544 75999-978 76000-546 75999-990 75999-870 75999-984 75999-982 75999-866 75999-988 75999-900 75999-986 75999-904 75999-868 75999-902 75999-908 75999-906 75999-880 75999-994 75999-874 75999-992 75999-872 75999-910 75999-998 75999-878 75999-996 75999-876 75999-914 75999-912 75999-918 75999-916 76000-726 76000-728 75999-892 75999-890 75999-884 75999-882 75999-888 75999-922 75999-886 75999-920 76000-730 75999-926 76000-732 75999-924 75999-928
FreeZone® 6 L Console Freeze Dryers, Labconco®
Freeze Dryers
FreeZone® 6 Liter Console Freeze Dryers are designed for lyophilizing moderate to large sample loads or numerous small container batches. Collectors are available to -50°C or -84°C.

  • Stay close to your samples as Lyo-Works™ operating system, provides mobile connectivity
  • Consistent, high quality results with flexible programming, and on board data collection
  • Challenging samples are easy with accessories designed specifically for laboratory samples
  • End-Zone™ End Point Detection option takes the guess work out of sample dryness

New Lyo-Works™ OS provides flexibility and convenience in an easy-to-use, touch screen display

The upright stainless steel collector coil holds 6 liters of ice before defrosting. Choice of a -50°C (for aqueous samples) or -84° C (for low eutectic point samples including acetonitrile) collector cooled by an environmentally-friendly HCFC/CFC-free refrigeration system. Internal coil maximizes vapor-trapping efficiency. Optional PTFE-coated collectors withstand corrosive compounds.

Lyo-Works OS provides a real time display of collector temperature and vacuum level on a 5" full color touch screen. Flexible programming and control of optional drying accessories including End-Zone™ end point detection for flask freeze drying. One-touch data logging and display provides information to maximize sample quality and minimize run time. Customizable settings include language, time zone, temperature and vacuum. Alerts and feedback of current operating parameters can be sent automatically to user-entered email addresses.

FreeZone exclusive features protect your vacuum pump from costly damage. A vacuum break valve bleeds air into the system when power to the freeze dryer or vacuum pump is shut off for more than 5 minutes. A moisture sensor prevents refrigeration or vacuum start-up when moisture is detected in the drain line. Vacuum pump is automatically shut down when a leak is present in initial pull down or during run. Front viewing window makes monitoring vacuum pump oil hassle free.

Your samples are protected with remote notification and data recording. Features include hot gas defrost, side-mounted USB port and rear-mounted Ethernet connection. The console provides interior space for a vacuum pump and accommodates the FreeZone Stoppering Tray Dryer (pump and tray dryer sold separately). These models offer the broadest range of built-in options including a shell freezer.

Certifications: 60Hz models conform to UL Standard 61010-1 and CAN/CSA C22.2 No. 61010-1. 50Hz models conform to CE Conformity marking.

Accessories information: Acrylic lid have 14.0" dia., with neoprene gasket and 3" dia. hole for direct connection of drying accessories to
collector. Includes cap for 3" dia. attachment port.

Ordering information: Vacuum pump with a displacement of at least 98 liters/minute, drying accessory and freeze dry glassware (if not bulk freeze drying) are required (not included).

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty