Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
FreeZone® 18 L Console Freeze Dryers with Stoppering Tray Dryer, Labconco®
Supplier: Labconco
FreeZone®
701821015 701822115 701821130 701821070 701821050 701821150 701822140 701821115 701822010 701822170 701822040 701822030 701821030 701822110 701822015 701822150 701822130 701822070 701821110 701821010 701821140 701821040 701821170 701822050
76000-618EA 77139.79 USD InStock
76000-618 76000-706 76000-708 76000-340 76000-330 76000-342 76000-332 76000-344 76000-322 76000-620 76000-334 76000-324 76000-622 76000-336 76000-710 76000-612 76000-326 76000-338 76000-712 76000-702 76000-328 76000-614 76000-704 76000-616
FreeZone® 18 L Console Freeze Dryers with Stoppering Tray Dryer, Labconco®
Freeze Dryers
FreeZone® 18 Liter Console Freeze Dryers with Stoppering Tray Dryers are designed for lyophilizing moderate to large loads and stoppering vials or serum bottles under vacuum.
  • Stay close to your samples as Lyo-Works™ operating system, provides mobile connectivity
  • Consistent, high quality results with flexible programming, and on board data collection
  • Challenging samples are easy with accessories designed specifically for laboratory samples
  • End-Zone™ End Point Detection option takes the guess work out of sample dryness
New Lyo-Works™ OS provides flexibility and convenience in an easy-to-use, touch screen display
The console’s upright stainless steel collector coil is capable of holding 18 liters of ice before defrosting. The -50° C (for aqueous samples) collector is cooled by an environmentally-friendly HCFC/CFC-free refrigeration system. Internal coil maximizes vapor-trapping efficiency. Optional PTFE-coated collectors withstand corrosive compounds.
Lyo-Works OS provides a real time display of collector temperature and vacuum level on a 5"" full color touch screen. Flexible programming and control of optional drying accessories including End-Zone™ end point detection for flask freeze drying. One-touch data logging and display provides information to maximize sample quality and minimize run time. Customizable settings include language, time zone, temperature and vacuum. Alerts and feedback of current operating parameters can be sent automatically to user-entered email addresses.
FreeZone exclusive features protect your vacuum pump from costly damage. A vacuum break valve bleeds air into the system when power to the freeze dryer or vacuum pump is shut off for more than 5 minutes. A moisture sensor prevents refrigeration or vacuum start-up when moisture is detected in the drain line. Vacuum pump is automatically shut down when a leak is present in initial pull down or during run. Front viewing window makes monitoring vacuum pump oil hassle free.
Your samples are protected with remote notification and data recording. Features include hot gas defrost, side-mounted USB port and rear-mounted Ethernet connection. The console provides interior space for a vacuum pump (sold separately).
Certifications: 60 Hz models conform to UL Standard 61010-1 and CAN/CSA C22.2 No. 61010-1. 50 Hz models conform to CE Conformity marking.
Accessories information: Stainless steel lid have 14.0" dia., with neoprene gasket and 3" dia. hole for direct connection of drying accessories to collector.
Includes cap for 3" dia. attachment port. Acrylic lid have 14.0" dia., with neoprene gasket and 3" dia. hole for direct connection of drying accessories to collector. Includes cap for 3" dia. attachment port.
Ordering information: Vacuum pump with a displacement of at least 163 liters/minute and freeze dry glassware (if not bulk freeze drying) are required (not included).
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty