Would you like to log in again?
SpillStopper™ Work Surfaces for Protector® ClassMate™ Laboratory Hoods, Labconco®
Supplier: Labconco
SpillStopper™
9600401 9600410 9600500 9600411 9600520 9600521 9600611 9600510 9600420 9600522 9600412 9600620 9600512 9600421 9600622 9600621 9600610 9600402 9600612 9600422 9600400 9600501 9600600 9600601 9600511 9600602 9600502
76045-940EA 4000 USD InStock
76045-940 76045-688 76045-666 76045-664 76045-686 76045-942 76045-668 76045-680 76045-684 76045-662 76045-660 76045-682 76045-938 76045-936 76045-676 76045-930 76045-674 76045-658 76045-934 76045-678 76045-932 76045-690 76045-672 76045-670 76045-926 76045-924 76045-928
SpillStopper™ Work Surfaces for Protector® ClassMate™ Laboratory Hoods, Labconco®
Fume Hoods and Enclosures Chemical Fume Hoods
Solid epoxy work surfaces are 3.2cm (11/4") thick, 65.7cm (257/8") deep, and dished to contain spills.
Solid epoxy cupsink cover (model 4005201; sold separately) covers cupsink when not in use.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty