Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
FreeZone® Triad® Benchtop Freeze Dryers, Labconco®
Supplier: Labconco
FreeZone®
794001070 794001010 794001040 794001030 794001015 794001050
76278-800EA 45940.73 USD InStock
76278-800 76278-798 76278-796 76278-838 76278-836 76278-834
FreeZone® Triad® Benchtop Freeze Dryers, Labconco®
Freeze Dryers
FreeZone® Triad® Benchtop Freeze Dryers are designed for multiple types of lyophilization: flask, bulk and stoppering vials. With the ability to freeze dry samples in so many different forms, the Triad’s versatility and compact design make it perfect for busy labs. Now with Lyo-Works™ OS the Triad is smarter than ever, making freeze drying even challenging samples easy.
  • Upright stainless steel collector coil capable of holding 2.5L of ice before defrosting
  • Lyo-Works™ OS offers expanded programs, real-time graphs and one-touch data collection on full color 5" capacitive touch screen display
  • Alerts can be emailed to user-entered email addresses for remote monitoring
This benchtop stoppering tray dryer and lyophilizer uses dual refrigeration systems in series to cool the collector coil to -85°C (-121°F), ideal for freeze drying samples with low eutectic points.
Lyo-Works™ OS provides a real time display of collector temperature and vacuum level on a 5"" full color, capacitive touch screen. Lyo-Works OS provides storage for up to 30 programs that can hold up to 16 steps each. Display includes navigation to start-up mode, vacuum set point, alerts, diagnostic tests, user manual, and defrost control. Data logging stores and displays data in table or chart format and data may be exported via the side-mounted USB or rear-mounted Ethernet connection. Language can be set to English, Spanish, French, German or Italian. Other settings include time, time zone, temperature (° C or ° F) and vacuum (mBar, Torr or Pa). Alerts and feedback of current operating programs can be sent automatically to user-entered email addresses for remote notification even when away from the lab.
The upright stainless steel collector coil holds 2.5 liters of ice before defrosting. Two 1/3 hp HCFC/CFC-free refrigeration compressors cool the collector to -85° C (-121° F), suitable for aqueous or dilute acetonitrile samples. Temperature-controlled shelves range from -55° C to +50° C (-67° F to +122° F) and a maximum of -75° C (-103° F) during pre-freezing. The shelves provide pneumatic stoppering with a diaphragm, eliminating the need for compressed gas. The space between the processing shelf and top of chamber accommodates containers with stoppers up to 148 mm maximum height. Other Triad features include a Pirani vacuum sensor and four valve ports for flasks.
FreeZone exclusive features protect the vacuum pump from damage. A vacuum break valve bleeds air into the system when power to the freeze dryer or vacuum pump is shut off for more than 5 minutes so moisture from the melting samples does not end up in the pump. If the vacuum level is higher than 5 mBar for 30 minutes, the vacuum pump will automatically turn off, preventing pump damage.
Certifications: Models conform to UL Standard 61010-1 (115/230V, 60 Hz models), CAN/CSA C22.2 No. 61010-1 (115/230V, 60 Hz models), and CE Conformity marking (230V, 50 Hz models).
Ordering information: Accessories not included. All models require (not included) a vacuum pump with a displacement of at least 98 L/min, 0.002 mBar ultimate pressure and fitting for 3/4" ID vacuum hose; and freeze dry glassware if not bulk freeze drying.
Order Now
ORDER
SPECIFICATIONS
SERVICES
Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty