Would you like to log in again?
RADIATION SURVEY METER FOR ANDROID OS
Supplier: S E INTERNATIONAL
Radiation Survey Meter For Android Os^ Radiation Survey Meter, Monitor 1000EC, is an energy compensated radiation detector that measures gamma, and x-rays, Users can choose from readings of CPM, CPS, uSv/hr, mR/hr, or in accumulated counts, It has red count light, beeper, an adjustable timer
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
RADIATION SURVEY METER FOR ANDROID OS 76299-466 MONITOR1000EC Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty