Would you like to log in again?
SNUFFER FIRE EXTINGUISHER 155 F 4X4X14IN
Supplier: Labconco
Snuffer Fire Extinguisher 155 F 4X4X14In^ Snuffer Fire Extinguisher, For use with 4/5ft Protector ClassMate Fume Hoods, use ABC dry powder to provide fire protection, fusible link melts at 155 F / chemical powder automatically discharges within 5 seconds to extinguish the fire, Dimensions: 4x4x14in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SNUFFER FIRE EXTINGUISHER 155 F 4X4X14IN 76308-694 1115006 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty