Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Purifier® Logic®+ Class II A2 Biosafety Cabinets, 230 V, India, Labconco
Supplier: Labconco
302481071 302310171 302510171 302480171 302581171 302680171 302410071 302580171 302480071 302381171 302511171 302411171 302311071 302481171 302681071 302611071 302681171 302381071 302510071 302581071 302311171 302380171 302610171 302610071 302411071 302611171 302680071 302580071 302310071 302410171 302380071 302511071
76319-436EA 20820 USD InStock
76319-436 76319-138 76319-478 76319-312 76319-356 76319-210 76319-232 76319-218 76319-186 76319-384 76319-260 76319-282 76319-268 76319-246 76319-422 76319-564 76319-520 76319-542 76319-464 76319-166 76319-342 76319-528 76319-506 76319-406 76319-328 76319-450 76319-296 76319-152 76319-570 76319-196 76319-492 76319-370
Purifier® Logic®+ Class II A2 Biosafety Cabinets, 230 V, India, Labconco
Fume Hoods and Enclosures Biological Safety Cabinets
Purifier Logic+ Type A2 biosafety cabinets combine ergonomics, comfort and safety.
  • MyLogic OS displays cabinet status and alarms with clear, concise images and messaging to build user confidence
  • Constant Airflow Profile™ (CAP) Technology automatically maintains a precise volume of air as the filters load to protect a user and their samples
  • Ergonomic Inclination™ Technology Suite includes 10° angled sash, patented curved air grille with built-in armrest, fully accessible controls and eye-level mounted color display
  • Quiet, efficient ECM blower and energy saving LED lights reduces cabinet’s carbon footprint
  • Provides personnel, product and environmental protection and is suitable for work in all biosafety levels
The Purifier Logic+ Class II A2 Biosafety Cabinet excels in user comfort and functionality. Decades of industry leading advances in ergonomic design have shaped the Inclination™ Technology Suite that includes the following: 10° angled sash, LED lighting, patented curved air grille with built in armrest, fully accessible controls and line-of-sight mounted color display with MyLogic™ operating system.
MyLogic OS provides intuitive control of the Logic+ with clear messages and vibrant imagery, status updates and alarms with diagnostic troubleshooting. Using ECM blower technology, the Logic+ uses the Constant Airflow Profile™ (CAP) to maintain a precise volume of air as the filters load. In addition, the cabinet will continuously display the HEPA filter life remaining and cabinet status. These features combine to provide unparalleled safety.
Type A2 biosafety cabinets provide personnel, product and environmental protection and are suitable for work in all biosafety levels. These biosafety cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through an efficient Ventus™ Canopy Connection (optional). In addition, some models have base stands and/or an Accessory Package (UV light, service fixture(s) and Vacu-Pass™ Cord and Cable Portal) included.
Certifications: NSF listed. ADA-compliant (height of controls and receptacles). CE Conformity marking (230 V models). ISO 5 conditions per ISO 14644-1 and 2.
Accessories information: Accessories not included unless stated.
Ordering information: Base stand required if model with stand is not selected.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty