Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Purifier® Logic®+ Class II A2 Biosafety Cabinets, 230 V Schuko, Labconco
Supplier: Labconco
302480031 302310031 302611131 302511131 302410031 302511031 302581131 302481131 302581031 302381131 302480131 302380131 302580131 302311131 302510131 302510031 302310131 302381031 302681031 302611031 302680131 302610131 302411031 302410131 302380031 302681131 302311031 302580031 302680031 302411131 302610031 302481031
76319-558EA 17680 USD InStock
76319-558 76319-514 76319-536 76319-458 76319-336 76319-554 76319-378 76319-430 76319-254 76319-276 76319-416 76319-240 76319-160 76319-180 76319-546 76319-304 76319-226 76319-204 76319-500 76319-400 76319-444 76319-146 76319-322 76319-486 76319-364 76319-132 76319-472 76319-174 76319-350 76319-392 76319-192 76319-290
Purifier® Logic®+ Class II A2 Biosafety Cabinets, 230 V Schuko, Labconco
Fume Hoods and Enclosures Biological Safety Cabinets
Purifier Logic®+ Type A2 biosafety cabinets combine ergonomics, comfort and safety.
  • MyLogic OS displays cabinet status and alarms with clear, concise images and messaging to build user confidence
  • Constant Airflow Profile™ (CAP) Technology automatically maintains a precise volume of air as the filters load to protect a user and their samples
  • Ergonomic Inclination™ Technology Suite includes 10° angled sash, patented curved air grille with built-in armrest, fully accessible controls and eye-level mounted color display
  • Quiet, efficient ECM blower and energy saving LED lights reduces cabinet’s carbon footprint
  • Provides personnel, product and environmental protection and is suitable for work in all biosafety levels
The Purifier Logic+ Class II A2 Biosafety Cabinet excels in user comfort and functionality. Decades of industry leading advances in ergonomic design have shaped the Inclination™ Technology Suite that includes the following: 10° angled sash, LED lighting, patented curved air grille with built in armrest, fully accessible controls and line-of-sight mounted color display with MyLogic™ operating system.
MyLogic OS provides intuitive control of the Logic+ with clear messages and vibrant imagery, status updates and alarms with diagnostic troubleshooting. Using ECM blower technology, the Logic+ uses the Constant Airflow Profile™ (CAP) to maintain a precise volume of air as the filters load. In addition, the cabinet will continuously display the HEPA filter life remaining and cabinet status. These features combine to provide unparalleled safety.
Type A2 biosafety cabinets provide personnel, product and environmental protection and are suitable for work in all biosafety levels. These biosafety cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through an efficient Ventus™ Canopy Connection (optional). In addition, some models have base stands and/or an Accessory Package (UV light, service fixture(s) and Vacu-Pass™ Cord and Cable Portal) included.
Certifications: NSF listed. ADA-compliant (height of controls and receptacles). CE Conformity marking (230 V models). ISO 5 conditions per ISO 14644-1 and 2.
Accessories information: Accessories not included unless stated.
Ordering information: Base stand required if model with stand is not selected.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty