Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Purifier® Cell Logic®+ Class II A2 Biosafety Cabinets, Labconco
Supplier: Labconco
342591001 342391001 342591101 332491101 342591011 322391011 322591111 332391101 342691111 322691111 342391101 322591001 332591111 332691111 332491001 342691101 332591001 332391001 332591101 322391111 342591111 322391101 322591011 332591011 332391011 332491011 332391111 342391011 322691011 342391111 332691101 322591101 322491011 342491111 342691001 332491111 332691001 342491001 322691101 322491111 342491101 332691011 342691011 322491101 342491011 322391001 322691001 322491001
76321-092EA 18818.1 USD InStock
76321-092 76321-090 76321-308 76321-258 76321-236 76321-136 76321-256 76321-234 76321-310 76321-218 76321-216 76321-272 76321-294 76321-150 76321-270 76321-292 76321-078 76321-198 76321-176 76321-076 76321-350 76321-148 76321-104 76321-324 76321-368 76321-126 76321-048 76321-224 76321-322 76321-200 76321-244 76321-046 76321-348 76321-106 76321-128 76321-226 76321-184 76321-162 76321-382 76321-062 76321-364 76321-242 76321-384 76321-186 76321-164 76321-362 76321-284 76321-064
Purifier® Cell Logic®+ Class II A2 Biosafety Cabinets, Labconco
Fume Hoods and Enclosures Biological Safety Cabinets
Purifier Cell Logic+ Type A2 biosafety cabinets combine ergonomics, comfort and safety.
  • MyLogic OS displays cabinet status and alarms with clear, concise images and messaging to build user confidence
  • Constant Airflow Profile™ (CAP) Technology automatically maintains a precise volume of air as the filters load to protect a user and their samples
  • Ergonomic Inclination™ Technology Suite includes 10° angled sash, patented curved air grille with built-in armrest, fully accessible controls and eye-level mounted color display
  • Quiet, efficient ECM blower and energy saving LED lights reduces cabinet’s carbon footprint
  • Provides personnel, product and environmental protection and is suitable for work in all biosafety levels
  • Specially designed for use with microscopes and/or temperature-sensitive samples
The Purifier Cell Logic+ Class II A2 Biosafety Cabinet excels in user comfort and functionality. Decades of industry leading advances in ergonomic design have shaped the Inclination™ Technology Suite that includes the following: 10° angled sash, LED lighting, patented curved air grille with built in armrest, fully accessible controls and line-of-sight mounted color display with MyLogic™ operating system.
MyLogic OS provides intuitive control of the Logic+ with clear messages and vibrant imagery, status updates and alarms with diagnostic troubleshooting. Using ECM blower technology, the Cell Logic+ uses the Constant Airflow Profile™ (CAP) to maintain a precise volume of air as the filters load. In addition, the cabinet will continuously display the HEPA filter life remaining and cabinet status. These features combine to provide unparalleled safety.
Cell Logic+ A2 Biosafety Cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through an efficient Ventus™ Canopy Connection (optional). Cell Logic+ models are also equipped to meet the specific needs of the cell culture industry. Optional features include the Scope-Ready™ and Temp-Zone™ packages. The Scope-Ready package allows the user’s microscope to be integrated into the biosafety cabinet. The Temp-Zone package includes a temperature controlled area on the work surface. The Cell Logic+ is available with the Scope-Ready package, the Temp-Zone package or both. Some cabinets include base stands and/or an Accessory Package (UV light (not on Scope-Ready models), service fixture(s) and Vacu-Pass™ Cord and Cable Portal).
Certifications: ETL listed. ADA-compliant (height of controls and receptacles). NSF and modified ASHRAE 110 compliant. CE Conformity marking (230 V models). ISO 5 conditions per ISO 14644-1 and 2.
Ordering information: Base stand required if model with stand is not selected. Microscope not included. Accessories not included unless stated.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty