Purifier Cell Logic+ Type B2 biosafety cabinets combine ergonomics, comfort and safety.
- MyLogic OS displays cabinet status and alarms with clear, concise images and messaging to build user confidence
- Constant Airflow Profile™ (CAP) Technology automatically maintains a precise volume of air as the filters load to protect a user and their samples
- Ergonomic Inclination™ Technology Suite includes 10° angled sash, patented curved air grille with built-in armrest, fully accessible controls and eye-level mounted color display
- Quiet, efficient ECM blower and energy saving LED lights reduces cabinet’s carbon footprint
- 100% exhaust design provides personnel, product and environmental protection and is suitable for work in all biosafety levels
- Specially designed for use with microscopes and/or temperature-sensitive samples
The Purifier Cell Logic+ Class II B2 Biosafety Cabinet excels in user comfort and functionality. Decades of industry leading advances in ergonomic design have shaped the Inclination™ Technology Suite that includes the following: 10° angled sash, LED lighting, patented curved air grille with built in armrest, fully accessible controls and line-of-sight mounted color display with MyLogic™ operating system.
MyLogic OS provides intuitive control of the Logic+ with clear messages and vibrant imagery, status updates and alarms with diagnostic troubleshooting. Using ECM blower technology, the Cell Logic+ uses the Constant Airflow Profile™ (CAP) to maintain a precise volume of air as the filters load. In addition, the cabinet will continuously display the HEPA filter life remaining and cabinet status. These features combine to provide unparalleled safety.
Cell Logic+ B2 Biosafety Cabinets exhaust 100% of their air and are designed to be hard-ducted to the outside. These biosafety cabinets are equipped to meet the specific needs of the cell culture industry. Optional features include the Scope-Ready™ and Temp-Zone™ packages. The Scope-Ready package allows the user’s microscope to be integrated into the biosafety cabinet. The Temp-Zone package includes a temperature controlled area on the work surface. The Cell Logic+ is available with the Scope-Ready package, the Temp-Zone package or both. Some cabinets include base stands and/or an Accessory Package (UV light (not on Scope-Ready models), service fixture(s) and Vacu-Pass™ Cord and Cable Portal).
Certifications: ETL listed (North America and Japan only). NSF compliant. CE Conformity marking (230V models). ISO 5 conditions per ISO 14644-1 and 2. ADA-Compliant (controls and receptacles).
Ordering information: Base stand required if model with stand is not selected. Accessories not included unless stated.