Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Purifier® Cell Logic®+ Class II B2 Biosafety Cabinets, Labconco
Supplier: Labconco
323481111 323681051 323481101 323681151 323481031 343481121 333681011 323681131 343681051 343681101 343481071 343481041 323481011 323681001 343681001 333481131 323481051 333681141 333681101 333481141 333481051 343481111 333481021 343681171 343681111 343681071 323681141 343681011 333481001 343681151 323681031 323681171 343681021 333681021 333481111 333681071 323481151 333681131 333681051 343481131 323481121 333481071 333481101 333681041 333681111 333681151 333681171 343681131 323681101 323481071 323681011 323481131 343681041 323481041 343481151 333481031 343481011 333681031 343481101 323681041 323681111 323481021 333681001 343481001 343681141 333481011 343481141 323681021 323481141 323681121 343481051 333481041 343681031 323481001 333481171 323481171 343481171 333481121 343681121 323681071 343481031 333481151 333681121 343481021
76321-508EA 29303.5 USD InStock
76321-508 76321-506 76321-428 76321-478 76321-434 76321-510 76321-554 76321-432 76321-476 76321-398 76321-552 76321-438 76321-558 76321-514 76321-436 76321-512 76321-556 76321-470 76321-474 76321-430 76321-396 76321-550 76321-472 76321-394 76321-418 76321-538 76321-544 76321-500 76321-466 76321-422 76321-542 76321-464 76321-420 76321-548 76321-504 76321-426 76321-502 76321-546 76321-468 76321-424 76321-540 76321-462 76321-460 76321-408 76321-528 76321-406 76321-412 76321-456 76321-532 76321-410 76321-498 76321-454 76321-530 76321-416 76321-536 76321-458 76321-414 76321-534 76321-492 76321-490 76321-452 76321-496 76321-494 76321-450 76321-518 76321-516 76321-522 76321-488 76321-400 76321-444 76321-520 76321-442 76321-486 76321-526 76321-404 76321-448 76321-524 76321-402 76321-446 76321-480 76321-484 76321-440 76321-560 76321-482
Purifier® Cell Logic®+ Class II B2 Biosafety Cabinets, Labconco
Fume Hoods and Enclosures Biological Safety Cabinets
Purifier Cell Logic+ Type B2 biosafety cabinets combine ergonomics, comfort and safety.
  • MyLogic OS displays cabinet status and alarms with clear, concise images and messaging to build user confidence
  • Constant Airflow Profile™ (CAP) Technology automatically maintains a precise volume of air as the filters load to protect a user and their samples
  • Ergonomic Inclination™ Technology Suite includes 10° angled sash, patented curved air grille with built-in armrest, fully accessible controls and eye-level mounted color display
  • Quiet, efficient ECM blower and energy saving LED lights reduces cabinet’s carbon footprint
  • 100% exhaust design provides personnel, product and environmental protection and is suitable for work in all biosafety levels
  • Specially designed for use with microscopes and/or temperature-sensitive samples
The Purifier Cell Logic+ Class II B2 Biosafety Cabinet excels in user comfort and functionality. Decades of industry leading advances in ergonomic design have shaped the Inclination™ Technology Suite that includes the following: 10° angled sash, LED lighting, patented curved air grille with built in armrest, fully accessible controls and line-of-sight mounted color display with MyLogic™ operating system.
MyLogic OS provides intuitive control of the Logic+ with clear messages and vibrant imagery, status updates and alarms with diagnostic troubleshooting. Using ECM blower technology, the Cell Logic+ uses the Constant Airflow Profile™ (CAP) to maintain a precise volume of air as the filters load. In addition, the cabinet will continuously display the HEPA filter life remaining and cabinet status. These features combine to provide unparalleled safety.
Cell Logic+ B2 Biosafety Cabinets exhaust 100% of their air and are designed to be hard-ducted to the outside. These biosafety cabinets are equipped to meet the specific needs of the cell culture industry. Optional features include the Scope-Ready™ and Temp-Zone™ packages. The Scope-Ready package allows the user’s microscope to be integrated into the biosafety cabinet. The Temp-Zone package includes a temperature controlled area on the work surface. The Cell Logic+ is available with the Scope-Ready package, the Temp-Zone package or both. Some cabinets include base stands and/or an Accessory Package (UV light (not on Scope-Ready models), service fixture(s) and Vacu-Pass™ Cord and Cable Portal).
Certifications: ETL listed (North America and Japan only). NSF compliant. CE Conformity marking (230V models). ISO 5 conditions per ISO 14644-1 and 2. ADA-Compliant (controls and receptacles).
Ordering information: Base stand required if model with stand is not selected. Accessories not included unless stated.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty