Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
PuriCare® Procedure Stations, 10" Sash, Labconco®
Supplier: Labconco
PuriCare®
312410011 312611051 312411031 312410041 312410001 312611021 312410071 312411071 312410021 312611031 312610011 312410031 312411041 312610041 312611041 312410051 312610071 312610021 312611001 312411051 312411001 312610031 312611071 312611011 312610001 312411021 312411011 312610051
76322-496EA 20035 USD InStock
76322-496 76322-474 76322-510 76322-476 76322-498 76322-478 76322-490 76322-492 76322-470 76322-472 76322-494 76323-362 76323-360 76322-504 76323-368 76322-506 76323-366 76322-508 76323-364 76322-486 76322-488 76322-468 76322-500 76322-502 76322-480 76322-482 76322-484 76323-370
PuriCare® Procedure Stations, 10" Sash, Labconco®
Fume Hoods and Enclosures Animal Research Enclosures
Specialized Class II A2 biological safety cabinets designed for conducting small animal research activities including cage changing, weighing, anesthesia delivery and surgical procedures.
  • MyLogic OS displays cabinet status and alarms with clear, concise images and messaging to build user confidence
  • Constant Airflow Profile™ (CAP) Technology automatically maintains a precise volume of air as the filters load to protect the user and their samples
  • Ergonomic Inclination™ Technology Suite includes 10° angled sash, curved air grille with built in armrest, fully accessible controls and eye-level mounted color display
  • Quiet, efficient ECM blower and energy saving LED lights reduces cabinet’s carbon footprint
  • Provides personnel, product and environmental protection and is suitable for work in all biosafety levels
PuriCare Procedure Stations are specialized Class II, Type A2 biosafety cabinets that create a particulate-free environment for conducting small animal research activities including cage changing, weighing, anesthesia delivery and surgical procedures. Advanced ergonomic design has shaped the Procedure Station’s Inclination™ Technology Suite that includes the following: 10° angled sash, LED lighting, patented curved air grille with built in armrest, fully accessible controls and line-of-sight mounted color display with MyLogic™ operating system.
MyLogic OS provides intuitive control of the Procedure Station with clear messages and vibrant imagery, status updates and alarms with diagnostic troubleshooting. Using ECM blower technology, the Procedure Station uses the Constant Airflow Profile™ (CAP) to maintain a precise volume of air as the filters load. In addition, the cabinet will continuously display the HEPA filter life remaining and cabinet status. These features combine to provide unparalleled safety.
Type A2 biosafety cabinets provide personnel, product and environmental protection and are suitable for work in all biosafety levels. These biosafety cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through an efficient Ventus™ Canopy Connection (optional). In addition, some models have base stands and/or an accessory package (254 nm UV light, ADA-compliant service fixture(s) with ball-type valve(s) and quarter turn handle, and NSF-approved Vacu-Pass™ Cord and Cable Portal) included.
Certifications: NSF International Listed; ETL listed; UL Standard 3101/61010-1; CAN/CSA C22.2 No. 1010.1-92; CE Conformity Marking (International models)
Ordering information: Base stand highly recommended (not included). Canopy (sold separately) may be required depending on operation.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty