Would you like to log in again?
TUBE ULTRA-CLEAR ROUND BTM 25X89MM BX 50
Supplier: BECKMAN COULTER
Tube Ultra-Clear Round Btm 25X89Mm BX50^ Tube, Ultra-Clear, round bottom, Closure: Open-Top Thinwall, Optical Property: Transparent, for use in Beckman Coulter instruments and rotors Type 70 Ti, Type 50.2 Ti, SW 32 Ti, SW 28, Nominal Capacity: 38 mL, Nominal Size: 25 x 89 mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBE ULTRA-CLEAR ROUND BTM 25X89MM BX 50 76335-350 344058 Box of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty