Would you like to log in again?
LINER POLYETHYLENE AIRFUGE 2.4 ML BX100
Supplier: BECKMAN COULTER
Liner Polyethylene Airfuge 2.4 Ml BX100^ Liner, Material: Polyethylene, Platform: Airfuge, for use with ACR-90 Chylomicron Rotor, Requires reusable adapter 342635, Nominal Capacity: 2.4 mL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LINER POLYETHYLENE AIRFUGE 2.4 ML BX100 76335-366 342634 Box of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty