Would you like to log in again?
TUBE THICKWALL POLYCARBONTE 7X20MM BX100
Supplier: BECKMAN COULTER
Tube Thickwall Polycarbonte 7X20Mm BX100^ Tube, Thickwall Polycarbonate, For use in Ultracentrifuge Rotors: Type 42.2 Ti, TLA-100, TLS-55, Closure: Open-Top, Nominal Capacity: 230 mL, Nominal Size: 7 x 20mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBE THICKWALL POLYCARBONTE 7X20MM BX100 76335-378 343775 Box of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty