Would you like to log in again?
TUBE ULTRA-CLEAR ROUND BTM 16X102MM BX50
Supplier: BECKMAN COULTER
Tube Ultra-Clear Round Btm 16X102Mm BX50^ Tube, Ultra-Clear, round bottom, Closure: Open-Top Thinwall, Optical Property: Transparent, allowing for easy location of visibly banded or pelleted samples, for use in SW 32.1 Ti, SW 28.1, Nominal Capacity: 17mL, Nominal Size: 16 x 102 mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBE ULTRA-CLEAR ROUND BTM 16X102MM BX50 76335-418 344061 Box of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty