Would you like to log in again?
TUBE THINWALL POLYPROPYLENE 4 ML BX50
Supplier: BECKMAN COULTER
Tube Thinwall Polypropylene 4 Ml BX50^ Tube, round bottom tube, Thinwall, Polypropylene, Closure: Open-Top, Optical Property: Transparent, designed for use in Beckman Coulter instruments and in SW 60 Ti rotor, Nominal Capacity: 4 mL, Nominal Size: 11 x 60 mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBE THINWALL POLYPROPYLENE 4 ML BX50 76335-422 328874 Box of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty