Would you like to log in again?
KIT TUBE FLOOR PREP 108 ML FOR 11.2 ML
Supplier: BECKMAN COULTER
Kit Tube Floor Prep 108 Ml For 11.2 Ml^ OptiSeal* Tube Kit, Polypropylene, Platform: Floor Prep, 11.2 mL For Use In Ultracentrifuge Rotors: NVT 65, Nominal Capacity: 108 mL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
KIT TUBE FLOOR PREP 108 ML FOR 11.2 ML 76335-554 360973 Kit of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty