Would you like to log in again?
TUBE THICKWALL POLYPROPYLENE 6 ML BX25
Supplier: BECKMAN COULTER
Tube Thickwall Polypropylene 6 Ml BX25^ Polypropylene Tube, Platform: Floor Prep, Closure: Open-Top Thickwall, For use in Benchtop and Microcentrifuges Rotors: F1010, S0410, For use in Ultracentrifuge Rotors: MLA-80, Nominal Capacity: 6 mL, Nominal Size: 16 x 64mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBE THICKWALL POLYPROPYLENE 6 ML BX25 76335-564 355646 Box of 25 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty