Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
TUBE POLYCARBONATE ROUND BTM 81ML BX25
Supplier: BECKMAN COULTER
Tube Polycarbonate Round Btm 81Ml BX25^ Open Top Polycarbonate Round Bottom tube, Thickwall, Platform: Floor Prep, Optical Property: Transparent, For use in Type 45 Ti rotor, Nominal Size: 38 x 102mm, Nominal Capacity: 81mL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBE POLYCARBONATE ROUND BTM 81ML BX25 76335-660 355628 Box of 25 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty