Would you like to log in again?
BOTTLE ASSEMBLY POLYCARBONATE 26 ML BX6
Supplier: BECKMAN COULTER
Bottle Assembly Polycarbonate 26 Ml BX6^ Polycarbonate Bottle with cap assembly, Platform: Floor Prep, used in benchtop centrifuge and microcentrifuge rotors F0630, Nominal Capacity: 26 mL, Nominal Size: 25 x 89 mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BOTTLE ASSEMBLY POLYCARBONATE 26 ML BX6 76335-676 355616 Box of 6 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty