Would you like to log in again?
Glove Box Gloves, Labconco
Supplier: Labconco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
5033100 5006300 5005501 5005600 5005502 5005500 5006000 5005601 5006200 5006100 5005800 5005900 5005602
32948-203PK 395 USD InStock
32948-203 32948-201 76344-358 32948-154 32948-110 76344-364 76344-362 76344-360 15202-519 32948-205 32948-019 32948-096 10790-744
Glove Box Gloves, Labconco
Gloves
Neoprene straight and accordion sleeves can be used with hand-specific neoprene or latex hands.
  • Hypalon® gloves are resistant to abrasion, ozone, and oxidizing chemicals
Neoprene, butyl, and Hypalon® gloves are one-piece, seamless, and hand-specific. Butyl gloves provide high impermeability to water vapor, gases, and toxic chemicals, and are resistant to oxygenated solvents and most oxidizing chemicals.
Ordering information: Sleeves come complete with O-rings and spring. Neoprene, butyl, and Hypalon® gloves come with O-rings.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty