Fully-featured, energy-efficient fume hood provides an environmentally-friendly solution to ducted fume hoods while simplifying installation.
- Fully featured fume hood
- Ducting is not required
- Energy efficient
- Environmentally friendly
- Uses Erlab's Neutrodine Unisorb filtration
- Four filter combinations available
- Models with no windows or back windows also available
The Protector® Echo™ Filtered Fume Hood combines the trusted filtration of Erlab’s Neutrodine® Unisorb Filter with the quality, innovation and energy-efficient fume hood designs of Labconco's unparalleled engineering to create an adaptable fully-featured fume hood that does not require ducting for easy installation. The Protector® Echo™ Filtered Fume Hood has a by-pass design and features many defining characteristics of Labconco Protector Hoods including the Eco-Foil™ Air Foil with Clean-Sweep openings, fiberglass-reinforced panel liner, fully closing sash, electrical receptacles and service fixtures.
The Neutrodine Unisorb Filter has been tested and found efficient on over 500 chemicals, and may last two years or more before requiring replacement. Because these filters allow the simultaneous handling of solvents, acids and bases, the Protector Echo may be used for a broad range of general chemistry fume hood applications. Plus, the use of these filters offers an eco-friendly solution to exhausting chemicals into the atmosphere. Options include acid or formaldehyde sensors or HEPA only for applications requiring particulate protection.
GFH Filtration Technology allows the Protector Echo Filtered Fume Hood to perform well beyond the SEFA 9-2010 DH III definition. User safety is maintained through numerous built-in features that constantly monitor and prevent unsafe conditions without relying on user practices or modifications. These safeguards include a sensor package that detects primary filter breakthrough of solvent and acid fumes, laboratory air quality, sash position and temperature, audio/visual alarms, and intelligent filter indication of type and status to prevent saturated filters being re-installed. The control box is conveniently located on the front of the hood and can be controlled by an app on the user’s phone.
Constructed with a glacier white powder-coated steel exterior and a chemical-resistant, fiberglass-reinforced, composite panel liner. The vertical-rising sash is a single piece of tempered safety glass with cable pulley and tracks with Clean-Sweep™ airflow openings. Sash stop is at 16" sash opening height. Sash opens to 28" high for loading. 37.5" (95.3 cm) high sightline from the work surface and header panel. Options include side and back windows for greater visibility and service fixtures.
Accessories information: Each hood requires a work surface, base cabinet or stand, and Neutrodine Unisorb Filters (sold separately). The number of filters needed depends on the hood's width. Optional accessories include HEPA Filter, prefilters, and Sash Opening Reduction Wing Kits.
Ordering information: The Protector Echo benchtop Filtered Fume Hood is available in three styles: no windows, side windows, and 360° windows, and four widths: 4', 5', 6', and 8'.
Caution: The Lifetime Payback Questionnaire (LPQ) form is required for all Protector Echo Filtered Fume Hood orders. It must be completed in full, submitted, reviewed, and approved before an order can be processed. Lead times do not start until the LPQ is approved. Contact your sales representative or Labconco for more information or a copy of the form.