Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Protector® Echo™ Floor-Mounted Filtered Fume Hoods, 55.7" Depth, Labconco Corporation
Supplier: Labconco
Protector®
183837601 183437401 183537602 183437820 183637621 183637400 183437421 183437420 183437602 183637802 183637820 183537401 183837421 183837401 183637420 183437801 183837420 183437200 183637421 183837402 183837221 183637801 183837400 183837220 183437201 183837621 183537620 183637620 183537421 183437202 183637202 183837202 183837802 183537221 183837801 183437402 183637401 183537402 183637821 183437802 183537801 183637602 183837800 183637201 183537601 183537800 183837820 183837821 183437821 183537202 183537821 183437400 183537220 183837200 183537802 183537201 183837620 183537400 183637402 183437220 183837602 183537600 183537621 183637600 183437601 183637200 183537820 183537420 183437621 183837600 183837201 183437800 183637601 183437600 183637800 183637221 183637220 183437221 183437620 183537200
76419-108EA 46233.75 USD InStock
76419-108 76419-228 76419-104 76419-346 76419-224 76419-348 76419-106 76419-226 76419-342 76419-100 76419-220 76419-344 76419-102 76419-222 76419-230 76419-350 76418-978 76418-976 76418-974 76418-972 76418-970 76419-118 76419-236 76419-356 76419-114 76419-238 76419-358 76419-116 76419-232 76419-110 76419-352 76419-234 76419-354 76419-112 76419-360 76419-362 76419-120 76419-240 76419-086 76418-968 76418-966 76419-328 76419-206 76419-208 76419-004 76419-326 76419-364 76419-122 76419-242 76419-000 76419-088 76419-124 76419-002 76419-244 76419-096 76419-098 76419-092 76419-094 76418-998 76419-090 76418-996 76418-994 76418-992 76418-990 76419-218 76419-338 76419-214 76419-334 76419-216 76419-336 76419-210 76419-330 76419-212 76419-332 76419-340 76418-988 76418-986 76418-984 76418-982 76418-980
Protector® Echo™ Floor-Mounted Filtered Fume Hoods, 55.7" Depth, Labconco Corporation
Fume Hoods and Enclosures Ductless Fume Hoods
Fully-featured, energy-efficient fume hoods provide an environmentally-friendly solution to ducted fume hoods while simplifying installation. Ideal for oversized equipment.
  • Fully featured fume hood
  • Ducting is not required
  • Energy efficient
  • Environmentally friendly
  • Uses Erlab's Neutrodine Unisorb filtration
  • Four filter combinations available
  • 37.7" and 43.7" (958 and 1110 mm) depths are also available
The Protector® Echo™ Floor-Mounted Filtered Fume Hood combines the trusted filtration of Erlab’s Neutrodine® Unisorb Filter with the quality, innovation and energy-efficient fume hood designs of Labconco's unparalleled engineering to create an adaptable fully-featured floor-mount fume hood that that does not require ducting for easy installation. The Protector® Echo™ Floor-Mounted Filtered Fume Hood has a by-pass design and features many defining characteristics of Labconco Protector Hoods including the Eco-Foil™ Air Foil with Clean-Sweep openings, fiberglass-reinforced panel liner, fully closing sash, electrical receptacles and service fixtures.
The Neutrodine Unisorb Filter has been tested and found efficient on over 500 chemicals, and may last two years or more before requiring replacement. Because these filters allow the simultaneous handling of solvents, acids and bases, the Protector Echo may be used for a broad range of general chemistry fume hood applications. Plus, the use of these filters offers an eco-friendly solution to exhausting chemicals into the atmosphere. Options include acid or formaldehyde sensors or HEPA only for applications requiring particulate protection.
GFH Filtration Technology allows the Protector Echo Floor-Mounted Filtered Fume Hood to perform well beyond the SEFA 9-2010 DH III definition. User safety is maintained through numerous built-in features that constantly monitor and prevent unsafe conditions without relying on user practices or modifications. These safeguards include a sensor package that detects primary filter breakthrough of solvent and acid fumes, laboratory air quality, sash position and temperature, audio/visual alarms, and intelligent filter indication of type and status to prevent saturated filters being re-installed. The control box is conveniently located on the front of the hood and can be controlled by an app on the user’s phone.
Constructed with a glacier white powder-coated steel exterior and a chemical-resistant, fiberglass-reinforced, composite panel liner. Two tempered safety glass vertical-rising sashes with powder-coated aluminum sash handles and powder-coated steel sash tracks with Clean-Sweep slots. Upper sash handle also includes Clean-Sweep openings. Sash stop is 52" from the floor. 73.5" (187 cm) high sightline from the floor to the header panel and 63.6" (162 cm) loading height.
Accessories information: Each hood requires Neutrodine Unisorb Filters (sold separately). The number of filters needed depends on the hood's width. Optional accessories include HEPA Filter and prefilters.
Ordering information: The Protector Echo Floor-Mounted Filtered Fume Hood is available in three exterior depths: 37.7", 43.7" and 55.7".
Caution: The Lifetime Payback Questionnaire (LPQ) form is required for all Protector Echo Filtered Fume Hood orders. It must be completed in full, submitted, reviewed, and approved before an order can be processed. Lead times do not start until the LPQ is approved. Contact your sales representative or Labconco for more information or a copy of the form.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty