Would you like to log in again?
Echo® and Airo® Filtered Fume Hood Accessories, Labconco
Supplier: Labconco
9577434 9577420 9577422 9577433 9577424 9577423 9577421
76436-354EA 1442 USD InStock
76436-354 76436-344 76436-356 76436-350 76436-352 76436-346 76436-348
Echo® and Airo® Filtered Fume Hood Accessories, Labconco
Fume Hoods and Enclosures Ductless Fume Hoods
Required and replacement accessories for Protector filtered fume hoods. These accessories are for use with Labconco Protector Echo® and Airo® filtered fume hoods. For all filters and prefilters: The number of filters/prefilters required depends on the width of the hood and if it is an initial order or a replacement order.
Neutrodine Unisorb molecular carbon filter for multi-purpose use with acids/bases/solvents, separated or combined. Compatible with over 700 chemicals.
HEPA filter, 99.995% efficient, H14 type.
Replacement roughing prefilter, located in light box.
Replacement acid roughing prefilter, located in light box. Treated for use with acids.
Replacement ammonia roughing prefilter, located in light box. Treated for use with ammonia.
Acid gas sensor is required for proper hood function on models with acid sensor. Replacement is required after hood alarm alerts user that shelf-life has been met (approximately 2 years).
Formaldehyde gas sensor is required for proper hood function on models with formaldehyde sensor. Replacement is required after hood alarm alerts user that shelf-life has been met (approximately 2 years).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty