Would you like to log in again?
Purifier® Logic+® and Axiom® Biosafety Cabinet Accessories, Labconco
Supplier: Labconco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Purifier®
3430501 7034803 3458401 3620301 3456101 7034801 3435602 3620302 7034800 3456103 3435603 3458403 3858320 3456102 3435601 3430500 7034802 3456100 3435600 3620303
76473-144EA 1030 USD InStock
76473-144 76473-122 76473-154 76473-132 76473-146 76473-124 76473-134 76473-156 76473-140 76473-150 76473-120 76473-142 76473-152 76473-130 76473-118 76473-148 76473-126 76473-136 76473-128 76473-138
Purifier® Logic+® and Axiom® Biosafety Cabinet Accessories, Labconco
Fume Hoods and Enclosures Biological Safety Cabinets
Accessories for use with Labconco Class II biosafety cabinets.
  • Accessories for various needs
  • Trace odor filters for nuisance odors
  • Prop rod holds work surface in raised position
BSC sash stop restricts the sash from closing to maintain continuous operation. For use with Labconco Class II biosafety cabinets. The dry contact relay kit for use with L3 generation Logic+ and Axiom biosafety cabinets.
Easy-clean prop rod holds the work surface in a raised position for easy cleaning of the sump area on Logic+ BSCs. Short version for use with service-fixtured BSCs. Long version for use with non-fixtured BSCs.
Replacement trace odor filter for Logic+ Type A2 trace odor filter kits. For use with Logic+ Type A2 biosafety cabinets. Trace odor filter kit for Logic+ A2 biosafety cabinets. Removes trace nuisance odors from the exhaust air of Logic+ Class II, Type A2 BSCs. Contact us for applications involving light-duty chemicals.
BSC flex duct for connection of ductwork to BSC exhaust damper. 10" Nominal diameter. Includes flex duct length and 2× stainless steel band clamps.
Axiom flat work surface. Flat replacement for standard dished Chem-Zone work surface. For use with Axiom Class II Type C1 biosafety cabinets.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty