Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Purifier® Axiom® Class II Type C1 Biosafety Cabinets, Labconco
Supplier: Labconco
Purifier®
304611171 304681171 304611101 304680141 304411041 304680041 304610131 304681121 304610051 304481151 304610111 304481021 304681141 304680101 304411111 304411151 304481071 304611111 304410071 304410131 304410111 304410121 304611141 304681001 304411071 304680171 304611071 304481031 304481111 304411021 304611121 304480021 304680071 304680121 304681021 304681031 304411121 304681111 304680151 304611011 304410101 304481131 304610171 304410171 304480101 304480131 304610031 304680111 304410141 304681101 304480171 304681151 304411101 304611051 304411031 304480031 304680131 304611001 304411141 304611041 304680021 304610071 304481011 304680011 304681071 304480071 304680031 304410031 304681011 304480051 304481141 304480121 304481171 304610151 304481051 304480001 304610011 304681051 304480041 304610141 304410001 304411171 304610041 304410041 304681131 304611031 304411001 304481041 304411051 304481121 304411011 304410051 304680001 304610021 304411131 304680051 304410021 304610001 304410151 304481001 304481101 304611151 304480151 304611131 304480141 304610121 304480011 304480111 304611021 304610101 304410011 304681041
76473-320EA 24720 USD InStock
76473-320 76473-166 76473-286 76473-322 76473-168 76473-288 76473-200 76473-162 76473-282 76473-164 76473-284 76473-328 76473-206 76473-208 76473-324 76473-202 76473-326 76473-204 76473-160 76473-280 76473-276 76473-278 76473-310 76473-272 76473-274 76473-316 76473-318 76473-312 76473-158 76473-314 76473-270 76473-342 76473-188 76473-220 76473-344 76473-222 76473-184 76473-186 76473-340 76473-228 76473-346 76473-224 76473-348 76473-226 76473-180 76473-182 76473-210 76473-298 76473-176 76473-330 76473-212 76473-178 76473-332 76473-294 76473-172 76473-296 76473-174 76473-218 76473-338 76473-214 76473-334 76473-216 76473-336 76473-290 76473-292 76473-170 76473-364 76473-242 76473-366 76473-244 76473-360 76473-362 76473-240 76473-368 76473-246 76473-248 76473-232 76473-198 76473-352 76473-234 76473-354 76473-194 76473-230 76473-350 76473-196 76473-236 76473-356 76473-238 76473-358 76473-190 76473-192 76473-264 76473-300 76473-266 76473-260 76473-262 76473-306 76473-308 76473-302 76473-268 76473-304 76473-380 76473-254 76473-374 76473-256 76473-376 76473-250 76473-370 76473-252 76473-372 76473-258 76473-378
Purifier® Axiom® Class II Type C1 Biosafety Cabinets, Labconco
Fume Hoods and Enclosures Biological Safety Cabinets
Flexible BSCs can be operated in Type A or B mode and changed as application needs change.
  • NSF Listed
  • Only Type C1 on the market
  • Maximum safety and flexibility
The Axiom Class II Type C1 maximizes safety and flexibility as the first biosafety cabinet that can be used in a recirculating Type A mode for standard microbiological work or can be connected to an exhaust system to function in Type B mode for handling hazardous chemicals. The unique design includes the Chem-Zone™, providing chemical safety with a large, clearly defined area within the enclosure, bringing peace of mind when handling chemicals. Other features that make the Axiom safe and easy to operate include its ergonomic Inclination™ Technology and the intuitive MyLogic™ Operating System.
Axiom Type C1 BSCs provide personnel, product and environmental protection by filtering hazardous particulates and aerosols and are suitable for work in all biosafety levels. These biosafety cabinets discharge HEPA filtered exhaust air directly back to the laboratory, or into an exhaust system through an included connection. Axiom models are available with base stands and/or an accessory package (UV light, service fixture(s) and Vacu-Pass™ Cord and Cable Portal) included.
Certifications: NSF listed. ETL listed. 230V models are CE listed.
Accessories information: Supporting base required if base stand option is not selected.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty