Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Purifier® Logic®+ Class II A2 Biosafety Cabinets with 12" Sash, Labconco Corporation

Supplier: Labconco
302421041 302620011 302520021 302621001 302520071 302521021 302520151 302521001 302421171 302521071 302520171 302520001 302521121 302421151 302621111 302621101 302420041 302621031 302620111 302621171 302521041 302420141 302421121 302620071 302420171 302520011 302621121 302420011 302421021 302420131 302421141 302621041 302421111 302420031 302621011 302621131 302621021 302520131 302620151 302420021 302420111 302621141 302421131 302620051 302521051 302521011 302520121 302520051 302620131 302520101 302621071 302620171 302421031 302620121 302521171 302421101 302420071 302521101 302620001 302521151 302421011 302520141 302421071 302520111 302620031 302620101 302620141 302420121 302520031 302421001 302420051 302621151 302420101 302620021 302521141 302621051 302520041 302420001 302521111 302521031 302421051 302521131 302420151 302620041
76473-880EA 18320 USD InStock
76473-880 76473-882 76473-922 76473-888 76473-844 76473-800 76473-924 76473-846 76473-802 76473-840 76473-884 76473-920 76473-842 76473-886 76473-918 76473-838 76473-914 76473-916 76473-870 76473-792 76473-872 76473-794 76473-790 76473-878 76473-834 76473-910 76473-836 76473-912 76473-874 76473-830 76473-796 76473-950 76473-832 76473-876 76473-952 76473-798 76473-908 76473-948 76473-904 76473-826 76473-906 76473-828 76473-860 76473-900 76473-944 76473-822 76473-866 76473-946 76473-902 76473-868 76473-824 76473-940 76473-786 76473-862 76473-942 76473-788 76473-820 76473-864 76473-816 76473-936 76473-818 76473-938 76473-892 76473-894 76473-850 76473-890 76473-812 76473-856 76473-932 76473-858 76473-814 76473-934 76473-852 76473-896 76473-854 76473-898 76473-810 76473-930 76473-808 76473-926 76473-804 76473-848 76473-928 76473-806
Purifier® Logic®+ Class II A2 Biosafety Cabinets with 12" Sash, Labconco Corporation
Fume Hoods and Enclosures Biological Safety Cabinets
Purifier® Logic®+ Type A2 biosafety cabinets with large sash opening create a particulate-free environment for small animal research activities such as cage changing, weighing, anesthesia delivery and surgical procedures.

  • Product and personnel protection
  • Large 12" sash opening
  • Erogonomic design

Purifier® Logic®+ Class II A2 biosafety cabinets excel in safety, performance, and comfort. Their ergonomic design with the Inclination™ Technology Suite which includes the following: MyLogic™ operating system with eye-level color display, 10° angled sash, curved air grille with built in armrest, plus fully accessible controls, outlets, and fixtures.

MyLogic is easy to use, displaying the BSC’s status with clear messaging and graphics that notify users of filter life, alerts, and alarms. The energy-efficient ECM blower powered by Labconco’s exclusive Constant Airflow Profile™ (CAP) technology ensures safety and reliability at all times.

Logic+ BSCs provide personnel, product and environmental protection and are suitable for work in all biosafety levels. These biosafety cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through the optional Canopy Connection. In addition, some models have base stands and/or an Accessory Package (UV light, service fixture(s) and Vacu-Pass™ Cord and Cable Portal) included.

Certifications: NSF listed. ETL listed. CAN/CSA. 230 V models are CE listed.

Ordering information: Supporting base is required if base stand option is not selected.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty