Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

PuriCare® Procedure Stations with 12" Sash, Labconco®

Supplier: Labconco
PuriCare®
312521051 312521001 312421001 312521031 312520031 312421041 312620021 312520041 312620051 312621011 312521071 312620031 312421031 312620001 312420011 312620011 312421021 312520001 312621041 312621071 312420071 312421071 312420041 312420001 312620071 312520011 312420031 312520071 312420021 312521011 312621031 312621001 312420051 312620041 312421051 312521041 312520021 312621021 312520051 312621051 312421011 312521021
76473-980EA 16835 USD InStock
76473-980 76473-982 76473-960 76473-988 76473-966 76473-968 76473-962 76473-984 76473-964 76473-986 76474-030 76474-012 76473-958 76474-034 76474-010 76474-032 76474-016 76474-014 76474-036 76474-018 76473-990 76473-992 76473-970 76473-976 76473-954 76473-998 76473-956 76473-978 76473-972 76473-994 76473-996 76473-974 76474-020 76474-002 76474-024 76474-022 76474-000 76474-006 76474-028 76474-026 76474-004 76474-008
PuriCare® Procedure Stations with 12" Sash, Labconco®
Fume Hoods and Enclosures Animal Research Enclosures
PuriCare Procedure Stations with a large sash opening create a particulate-free environment for small animal research activities such as cage changing, weighing, anesthesia delivery and surgical procedures.

  • Product and personnel protection
  • Large 12" sash opening
  • Erogonomic design

PuriCare Procedure Stations are Class II A2 biosafety cabinets that excel in safety, performance, and comfort. Their ergonomic design with the Inclination™ Technology Suite which includes the following: MyLogic™ operating system with eye-level color display, 10° angled sash, curved air grille with built in armrest, plus fully accessible controls, outlets, and fixtures.

MyLogic is easy to use, displaying the BSC’s status with clear messaging and graphics that notify users of filter life, alerts, and alarms. The energy-efficient ECM blower powered by Labconco’s exclusive Constant Airflow Profile™ (CAP) technology ensures safety and reliability at all times.

Procedure Station BSCs provide personnel, product and environmental protection and are suitable for work in all biosafety levels. These biosafety cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through the optional Canopy Connection. They contain a robust pre-filter under the work surface to trap dander, hair, and other particulates from the supply and exhaust HEPA filters. In addition, some models have base stands and/or an accessory package (UV light, service fixture(s) and Vacu-Pass™ Cord and Cable Portal) included.

Certifications: NSF listed. ETL listed. CAN/CSA. 230 V models are CE listed.

Ordering information: Supporting base is required.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty