Would you like to log in again?
Spectrum Blowers
Supplier: Labconco
Spectrum
177250100 177051100 177050100 177251200 177252200 177052100 177252100 177250200 177251100
76645-808EA 8692.15 USD InStock
76645-808 76645-818 76645-806 76645-816 76645-814 76645-812 76645-822 76645-810 76645-820
Spectrum Blowers
Ventilators
Spectrum blowers with onboard intelligence that integrate with any application and building management system.
  • Performance range simplifies blower sizing
  • Exceptionally efficient and easy to maintain
  • Connects to almost any building management system
  • Streamlines set-up and remote monitoring
Energy efficiency is highlighted by the Electronically Commutated Motor (ECM) for 1 horsepower models and Variable Frequency Drive (VFD) for 2 horsepower models, both types consume less energy and are less expensive to maintain than traditional belt driven motors.
The PVC spectrum is recommended for laboratories that might use perchloric acid and would handle applications that require washdown and ducting.
The epoxy coated steel material is an economical option for laboratories using solvents or very mild acids.
The fiberglass spectrum is best for fume hood exhaust systems with corrosive conditions.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty